Thermo Scientific Chemicals

(S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid, 98%, 94% ee, Thermo Scientific Chemicals

Catalog number: 299142500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

(S)-(+)-2-Amino-3-hydroxy-3-methylbutanoic acid, 98%, 94% ee, Thermo Scientific Chemicals

Catalog number: 299142500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 299142500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2280-27-5
IUPAC Name
(2S)-2-amino-3-hydroxy-3-methylbutanoic acid
Molecular Formula
C5H11NO3
InChI Key
LDRFQSZFVGJGGP-GSVOUGTGSA-N
SMILES
CC(C)(O)[C@H](N)C(O)=O
TLC
>=97.5 %
Appearance (Form)
Powder
Specific optical rotation
+11° to +14° (20°C, 589 nm) (c=2, 6 N HCl)
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text