Amberlite™ XAD 1180 ion-exchange resin, Thermo Scientific Chemicals
Amberlite™ XAD 1180 ion-exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ XAD 1180 ion-exchange resin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic Bottle
Catalog number 299151000
Price (USD)/ Each
40.00
-
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
40.00
Add to cart
Amberlite™ XAD 1180 ion-exchange resin, Thermo Scientific Chemicals
Catalog number299151000
Price (USD)/ Each
40.00
-
Add to cart
Chemical Identifiers
CAS9003-69-4
MDL NumberMFCD00132704
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Moisture holding capacity61 to 67 %
Harmonic mean size0.350 to 0.600 mm
Particle size=<10.0 % (> 0.850 mm)
Uniformity coefficient=<1.80
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only