Thermo Scientific Chemicals

Amberlite™ XAD 1180 ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 299155000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Amberlite™ XAD 1180 ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 299155000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 299155000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
9003-69-4
MDL Number
MFCD00132704
Appearance (Color)
White to light yellow
Moisture holding capacity
61 to 67 %
Harmonic mean size
0.350 to 0.600 mm
Particle size
=<10.0 % (> 0.850 mm)
Uniformity coefficient
=<1.80

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text