Thermo Scientific™

Pierce™ Disposable Columns, 5 mL

Catalog number: 29922
Have Questions?
Thermo Scientific™

Pierce™ Disposable Columns, 5 mL

Catalog number: 29922
Have Questions?
Catalog Number
29922
Unit Size
100 columns
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
29922100 columns
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.

These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10 mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.

Features of these disposable plastic columns:

• For benchtop gravity-flow affinity purification and size exclusion chromatography
• Supplied complete with porous polyethylene discs, stoppers and end caps
• Columns can be pre-packed and stored until needed
• Compatible with most types of aqueous buffer eluents commonly used in chromatography
• Plastic funnels can be attached to expand the buffer reservoir for increased flow rates

Applications:
• Affinity purification/Affinity chromatography
• Immunodepletion
• Size-exclusion chromatography

Related Products
Pierce™ Disposable Column Trial Pack
Pierce™ Disposable Columns, 2 mL
Pierce™ Disposable Columns, 10 mL
Column Funnels for Pierce™ Disposable Columns
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
5 mL
Description
Disposable Columns, 5mL
Product Line
Pierce™
Quantity
100 columns
Type
Spin Column
Format
Gravity Flow Column
Column Type
Empty Column, Polyethylene Filter
Unit Size
100 columns

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text