Pierce™ Disposable Column Trial Pack
Product Images
Thermo Scientific™
Pierce™ Disposable Column Trial Pack
Thermo Scientific Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety ofRead more
Have Questions?
Catalog number 29925
Price (USD)/ 6 columns
111.00
-
Add to cart
Price (USD)/ 6 columns
111.00
Add to cart
Pierce™ Disposable Column Trial Pack
Catalog number29925
Price (USD)/ 6 columns
111.00
-
Add to cart
Thermo Scientific Pierce Disposable Plastic Columns are empty columns with matching frits and caps for use in a variety of resin-based affinity or desalting techniques.

These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10 mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.

Features of these disposable plastic columns:

• For benchtop gravity-flow affinity purification and size exclusion chromatography
• Supplied complete with porous polyethylene discs, stoppers and end caps
• Columns can be pre-packed and stored until needed
• Compatible with most types of aqueous buffer eluents commonly used in chromatography
• Plastic funnels can be attached to expand the buffer reservoir for increased flow rates

Applications:
• Affinity purification/Affinity chromatography
• Immunodepletion
• Size-exclusion chromatography

Related Products
Pierce™ Disposable Columns, 2 mL
Pierce™ Disposable Columns, 5 mL
Pierce™ Disposable Columns, 10 mL
Column Funnels for Pierce™ Disposable Columns
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionDisposable Column Trial Pack
Product LinePierce™
Quantity6 columns
TypeSpin Column
FormatGravity Flow Column
Column TypeEmpty Column
Unit Size6 columns
Contents & Storage
Store at room temperature.