Chemical Identifiers
CAS13965-03-2
IUPAC Namepalladium(2+) bis(triphenylphosphane) dichloride
Molecular FormulaC36H30Cl2P2Pd
InChI KeyYNHIGQDRGKUECZ-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more