Thermo Scientific Chemicals

1,3-Dioxolane, 99.5+%, pure, stabilized, Thermo Scientific Chemicals

Catalog number: 299480010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

1,3-Dioxolane, 99.5+%, pure, stabilized, Thermo Scientific Chemicals

Catalog number: 299480010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 299480010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
646-06-0
IUPAC Name
1,3-dioxolane
Molecular Formula
C3H6O2
InChI Key
WNXJIVFYUVYPPR-UHFFFAOYSA-N
SMILES
C1COCO1
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.3990 to 1.4010 (20°C, 589 nm)
Stabilizer
60 to 100 ppm BHT
GC
>=99.50 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text