Zirconium fluoride, 99.5%, Thermo Scientific Chemicals
Zirconium fluoride, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium fluoride, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 299791000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-64-4
IUPAC Namezirconium(4+) tetrafluoride
Molecular FormulaF4Zr
InChI KeyOMQSJNWFFJOIMO-UHFFFAOYSA-J
SMILES[F-].[F-].[F-].[F-].[Zr+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay>=99.4 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only