Thermo Scientific™

Pierce™ Recombinant Protein G, Biotinylated

Catalog number: 29988
Have Questions?
Thermo Scientific™

Pierce™ Recombinant Protein G, Biotinylated

Catalog number: 29988
Have Questions?
Catalog Number
29988
Unit Size
0.5 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
299880.5 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Use Thermo Scientific Pierce Biotinylated Recombinant Protein G to probe and detect mouse and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

Features of Biotinylated Recombinant Protein G:

• Contains two Fc-binding domains per protein
• Better than Protein A for mouse (incl. IgG1), human (incl. IgG3) and rat, goat and cow antibodies
• Poorer than Protein A for guinea pig, pig, dog and cat
• Does not bind human IgM, IgD or IgA

Protein G is a bacterial cell wall protein isolated from group G Streptococci. DNA sequencing of native Protein G identifies two IgG-binding domains and sites for albumin and cell surface binding. The albumin and cell surface binding domains have been eliminated from Recombinant Protein G to reduce nonspecific binding and, therefore, can be used to separate IgG from crude samples. Optimal binding occurs at pH 5, although binding is also effective at pH 7.0 to 7.2.

Properties of Recombinant Protein G:
• Source: E. coli
• Molecular Weight: ∼21,600 (Apparent MW by SDS-PAGE: 32,000)
• Form: salt-free powder
• A280 of 0.1% solution: 1.0
• Isoelectric point (pI): 4.5

Because Protein G has greater affinity than Protein A for most mammalian IgGs, it may be used for the purification of mammalian IgGs that do not bind well to Protein A. Protein G binds with significantly greater capacity than Protein A to several IgG subclasses such as human IgG3, mouse IgG1 and rat IgG2a. However, Protein G does not bind to human IgM, IgD and IgA. Differences in binding characteristics between Protein A and Protein G may be explained by the differing compositions in the IgG-binding sites of each protein. The tertiary structures of these proteins are very similar although their amino acid compositions are significantly different.

There are inconsistencies in reported binding properties of IgG to Protein G. Variations in isolation and manufacturing methods for Protein G may affect IgG binding, partially because there are differing numbers of IgG-binding sites on various sources of Protein G. Binding studies have been performed using native Protein G and several different recombinant forms. Several assay methods have been used to determine relative affinity, including radiolabeling experiments and ELISA techniques. The differing affinity assays may explain some of the inconsistencies. In addition, there are significant binding differences when different buffers are used. Approximately 44% more IgG from rat serum bound to Protein G when Protein G Binding Buffer was used as compared with 20 mM Tris Buffer, pH 7.5.

Related Products
Pierce™ Recombinant Protein G
Pierce™ Recombinant Protein G, Peroxidase Conjugated
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Protein G, Biotinylated
Conjugate
Biotin
Quantity
0.5 mg
Product Line
Pierce™
Form
Powder
Protein Subtype
Protein G
Target
Antibodies

Contents & Storage

Upon receipt store at -20°C. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text