Thermo Scientific™

Pierce™ Protein A, Biotinylated

Catalog number: 29989
Have Questions?
Thermo Scientific™

Pierce™ Protein A, Biotinylated

Catalog number: 29989
Have Questions?
Catalog Number
29989
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
299891 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Use this biotinylated form of Thermo Scientific Pierce Protein A to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

Features of Protein A:

• Contains four Fc-binding domains per protein
• Better than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
• Poorer than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies

Protein A is a cell wall component produced by several strains of Staphylococcus aureus that consists of a single polypeptide chain and contains little or no carbohydrate. The Protein A molecule contains four high-affinity binding sites capable of interacting with the Fc region from IgG of several species including human and rabbit. Optimal binding occurs at pH 8.2, although binding is also effective at neutral or physiological conditions (pH 7.0 to 7.6).

Properties of Native Protein A:
• Source: Staphylococcus aureus
• Molecular weight: 46,762 (Apparent MW by SDS-PAGE: 42,000)
• Form: salt-free powder
• A280 of 0.1% solution: ∼0.137 (theoretical)
• Isoelectric point (pI): 5.16 (theoretical)

The interaction between Protein A and IgG is not equivalent for all species. Even within a species, Protein A interacts with some subclasses of IgG and not others. For instance, human IgG1, IgG2 and IgG4 bind strongly, while IgG3 does not bind. There are also many instances in which monoclonal antibodies do not bind to Protein A, especially the majority of rat immunoglobulins and mouse IgG1.

Related Products
Pierce™ Protein A
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Protein A
Conjugate
Biotin
Quantity
1 mg
Product Line
Pierce™
Form
Powder
Protein Subtype
Protein A
Target
Antibodies

Contents & Storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text