1x1 image pixel for data collection
Nalgene™ Reusable Filter Units
Nalgene™ Reusable Filter Units
Thermo Scientific™
Nalgene™ Reusable Filter Units
Eliminate the need for a manifold or breakable glass vacuum flask. Nalgene Reusable Filter Holders with Receiver are complete, environmentally-friendly systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric) ReceiverUnit Size
300-4000250 mLCase of 4
300-4000PK
also known as 300-4000
250 mLEach
300-4050500 mLCase of 4
300-4050PK500 mLEach
300-41001000 mLCase of 4
300-4100PK1000 mLEach
Catalog number 300-4000
Price (USD)
505.40
Online Exclusive
758.00
Save 252.60 (33%)
Case of 4
Add to cart
Volume (Metric) Receiver:
250 mL
Unit Size:
Case of 4
Price (USD)
505.40
Online Exclusive
758.00
Save 252.60 (33%)
Case of 4
Add to cart
Reusable filter units include two membrane support plates: a clear one-piece plate for sterilization applications and a two-piece white plate for analytical applications. Accepts 47 or 50 mm diameter membranes. Two side-arms allow connection to vacuum line. Tubing adapters accept 1/4- to 5/16-inch (6 to 8 mm) I.D. vacuum tubing. 4 units per case.
Features
  • Replaces metal or breakable glass – major components are made of durable, transparent, autoclavable polysulfone
  • Use for sterilization or analysis. Filter holders come with sterilization and analytical membrane support plates
  • Designed to be used with vacuum or pressure
  • Graduated chambers for easy reference
Specifications
AutoclavableAutoclavable
No. per Pack1
Volume (Metric) Receiver250 mL
Volume (Metric) Upper250 mL
MaterialPolysulfone
Unit SizeCase of 4
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection