Rubidium nitrate, 99.8%, pure, Thermo Scientific Chemicals
Rubidium nitrate, 99.8%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rubidium nitrate, 99.8%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 300070100
Price (USD)/ Each
62.60
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
62.60
Add to cart
Rubidium nitrate, 99.8%, pure, Thermo Scientific Chemicals
Catalog number300070100
Price (USD)/ Each
62.60
-
Add to cart
Chemical Identifiers
IUPAC Namerubidium(1+) nitrate
Molecular FormulaNO3Rb
InChI KeyRTHYXYOJKHGZJT-UHFFFAOYSA-N
SMILES[Rb+].[O-][N+]([O-])=O
Molecular Weight (g/mol)147.47
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.2 %
Appearance (Form)Crystals or needles
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only