AppearanceMay develop some turbidity or precipitate
Titration Argentometric98.50 to 101.50 % (ex Cl)
Appearance (Form)Liquid
Refractive index1.4520 to 1.4560 (20°C, 589 nm)
Appearance (Color)Colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.