Calcium hypochlorite, tech., granular, 65% available chlorine
Calcium hypochlorite, tech., granular, 65% available chlorine
Calcium hypochlorite, tech., granular, 65% available chlorine
Calcium hypochlorite, tech., granular, 65% available chlorine
Thermo Scientific Chemicals

Calcium hypochlorite, tech., granular, 65% available chlorine

CAS: 7778-54-3 | CaCl2O2 | 142.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 300341000
Price (USD)
31.65
Special offer
Online exclusive
Ends: 26-Sep-2025
39.40
Save 7.75 (20%)
Each
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 26-Sep-2025
39.40
Save 7.75 (20%)
Each
Add to cart
Chemical Identifiers
CAS7778-54-3
IUPAC Namecalcium dihypochlorite
Molecular FormulaCaCl2O2
InChI KeyZKQDCIXGCQPQNV-UHFFFAOYSA-N
SMILES[Ca++].[O-]Cl.[O-]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Chlorine (Cl)72 % (Available chlorine) (typical)
Sodium chloride (NaCl)8.5 % (typical)
Calcium chloride (CaCl)1.0 % (typical)
Calcium chlorate (Ca(ClO3)2)1.0 % (typical)
Calcium hydroxide (Ca(OH)2)4.5 % (typical)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Share catalog number, name or link