4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent, Thermo Scientific Chemicals
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number 300601000
Price (USD)/ Each
315.65
Online exclusive
350.00 
Save 34.35 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
315.65
Online exclusive
350.00 
Save 34.35 (10%)
Add to cart
4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, ACS reagent, Thermo Scientific Chemicals
Catalog number300601000
Price (USD)/ Each
315.65
Online exclusive
350.00 
Save 34.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name4,5-dihydroxynaphthalene-2,7-disulfonate
Molecular FormulaC10H6O8S2
InChI KeyHLVXFWDLRHCZEI-UHFFFAOYSA-L
SMILESOC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol)318.27
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to grey to light brown
Infrared spectrumConforms
Sensitivity(to nitrate) Passes test
Sensitivity(to formaldehyde) Passes test
Clarity of solutionPasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only