Thermo Scientific Chemicals

Se-methylseleno-L-cysteine, 98%

Catalog number: 300640500
50 mg, Glass Bottle, Each
Thermo Scientific Chemicals

Se-methylseleno-L-cysteine, 98%

Catalog number: 300640500
50 mg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 300640500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26046-90-2
IUPAC Name
(2R)-2-amino-3-(methylselanyl)propanoic acid
Molecular Formula
C4H9NO2Se
InChI Key
XDSSPSLGNGIIHP-VKHMYHEASA-N
SMILES
C[Se]C[C@H](N)C(O)=O
Appearance (Form)
Powder
Titration with HClO4
>=97.5 %
Infrared spectrum
Conforms
Specific optical rotation
-13° to -15° (20°C, 589 nm) (c=1, water)
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text