Methanol-d3, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Methanol-d3, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol-d3, for NMR, 99.5% atom D, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
2.5 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 300730100
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
-
Add to cart
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
Add to cart
Methanol-d3, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Catalog number300730100
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1849-29-2
IUPAC Name(²H₃)methanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-FIBGUPNXSA-N
SMILES[2H]C([2H])([2H])O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
D-Enrichment>=99.4 %
Appearance (Form)Liquid
Water=<0.1 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only