Thermo Scientific Chemicals

Calcium bromide hydrate, for analysis, Thermo Scientific Chemicals

Catalog number: 300840010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium bromide hydrate, for analysis, Thermo Scientific Chemicals

Catalog number: 300840010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 300840010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
71626-99-8
IUPAC Name
calcium dibromide
Molecular Formula
Br2Ca
InChI Key
WGEFECGEFUFIQW-UHFFFAOYSA-L
SMILES
[Ca++].[Br-].[Br-]
Appearance (Color)
White
Titration Complexometric
>=83.0 % (CaBr2)
Barium (Ba)
Passes test
Sulfate (SO4)
=<100 ppm
Heavy metals (as Pb)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text