Chemical Identifiers
CAS10127-02-3
IUPAC Namezinc(2+) hydrogen N3,N3,N6,N6-tetramethylacridine-3,6-diamine trichloride
Molecular FormulaC17H20Cl3N3Zn
InChI KeyVADJQOXWNSPOQA-UHFFFAOYSA-L
SMILES[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
View more