Acridine Orange, pure, ca. 55% dye content
Acridine Orange, pure, ca. 55% dye content
Thermo Scientific Chemicals

Acridine Orange, pure, ca. 55% dye content

 
Have Questions?
Quantity:
100 g
Catalog number 300911000
Price (USD)/ Each
179.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
179.00
Add to cart
Acridine Orange, pure, ca. 55% dye content
Catalog number300911000
Price (USD)/ Each
179.00
-
Add to cart
Chemical Identifiers
CAS10127-02-3
IUPAC Namezinc(2+) hydrogen N3,N3,N6,N6-tetramethylacridine-3,6-diamine trichloride
Molecular FormulaC17H20Cl3N3Zn
InChI KeyVADJQOXWNSPOQA-UHFFFAOYSA-L
SMILES[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrumComplies
Infrared spectrumConforms
Dye content>=50 %
Appearance (Form)Powder
Appearance (Color)Red-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only