cis-1,2-Cyclohexanediol, 99%, Thermo Scientific Chemicals
cis-1,2-Cyclohexanediol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-1,2-Cyclohexanediol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 300970010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1792-81-0
IUPAC Namecyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-UHFFFAOYNA-N
SMILESOC1CCCCC1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Appearance (Color)White to light beige
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only