Thermo Scientific Chemicals

Amberjet™ 4200-Cl, Thermo Scientific Chemicals

Catalog number: 301335000
500 g, Each
Thermo Scientific Chemicals

Amberjet™ 4200-Cl, Thermo Scientific Chemicals

Catalog number: 301335000
500 g, Each
Quantity
Catalog number: 301335000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60177-39-1
Uniformity coefficient
=<1.25
Moisture holding capacity
49 to 55 % (Cl)
Particle size
=<0.5 % (< 0.425 mm)
Appearance (Color)
Semitransparent yellow
Harmonic mean size
0.600 to 0.800 mm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text