Thermo Scientific Chemicals

Amberlite™ HPR900, OH-form ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 301345000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Amberlite™ HPR900, OH-form ion-exchange resin, Thermo Scientific Chemicals

Catalog number: 301345000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 301345000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
197809-12-4
MDL Number
MFCD00802695
Chloride (Cl)
=<0.5 % (Chloride sites content)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text