1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 301420010
Price (USD)/ Each
277.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
327.00 
Save 49.35 (15%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
277.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
327.00 
Save 49.35 (15%)
Add to cart
1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals
Catalog number301420010
Price (USD)/ Each
277.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
327.00 
Save 49.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1-aminocyclopropane-1-carboxylic acid
Molecular FormulaC4H7NO2
InChI KeyPAJPWUMXBYXFCZ-UHFFFAOYSA-N
SMILESNC1(CC1)C(O)=O
Molecular Weight (g/mol)101.11
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<9 % (K.F.)
Appearance (Form)Powder and/or granules
Titration with NaOH>=98.5 % (On dry substance)
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only