Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 301422500
250 mg, Glass bottle, Each
Thermo Scientific Chemicals

1-Amino-1-cyclopropanecarboxylic acid, 99%, Thermo Scientific Chemicals

Catalog number: 301422500
250 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 301422500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
22059-21-8
IUPAC Name
1-aminocyclopropane-1-carboxylic acid
Molecular Formula
C4H7NO2
InChI Key
PAJPWUMXBYXFCZ-UHFFFAOYSA-N
SMILES
NC1(CC1)C(O)=O
Appearance (Form)
Powder and/or granules
Titration with NaOH
>=98.5 % (On dry substance)
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Water
=<9 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text