Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%

Catalog number: 301605000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, tripotassium salt dihydrate, 99+%

Catalog number: 301605000
500 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
65501-24-8
IUPAC Name
tripotassium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Formula
C10H17K3N2O10
InChI Key
MAPFUJCWRWFQIY-UHFFFAOYSA-K
SMILES
O.O.[K+].[K+].[K+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Appearance (Color)
White
Water
=<9 % (K.F.)
Appearance (Form)
Crystalline powder
Titration Complexometric
>=99.0 % (on dry substance)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text