Thermo Scientific Chemicals

Diphenyl ditelluride, 97%, Thermo Scientific Chemicals

Catalog number: 301840010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenyl ditelluride, 97%, Thermo Scientific Chemicals

Catalog number: 301840010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 301840010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32294-60-3
IUPAC Name
(phenylditellanyl)benzene
Molecular Formula
C12H10Te2
InChI Key
VRLFOXMNTSYGMX-UHFFFAOYSA-N
SMILES
[Te]([Te]C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Powder or crystals
Appearance (Color)
Orange to brown
Infrared spectrum
Conforms
HPLC
>=96.0 %
Melting point
64°C to 67°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text