Thermo Scientific Chemicals

Imidazole, 99+%, crystalline, Thermo Scientific Chemicals

Catalog number: 301870010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Imidazole, 99+%, crystalline, Thermo Scientific Chemicals

Catalog number: 301870010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 301870010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
288-32-4
IUPAC Name
1H-imidazole
Molecular Formula
C3H4N2
InChI Key
RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES
N1C=CN=C1
Appearance (Color)
White to light yellow to light beige
Infrared spectrum
Conforms
Titration with HClO4
>=99.0 % (On dry substance)
Appearance (Form)
Crystalline powder
Melting point
86°C to 90°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text