Imidazole, 99+%, crystalline, Thermo Scientific Chemicals
Imidazole, 99+%, crystalline, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Imidazole, 99+%, crystalline, Thermo Scientific Chemicals

Imidazole, 99+%, C3H4N2, CAS Number-288-32-4, glyoxalin, imidazole, miazole, imutex, pyrro b monazole, 1,3-diazole, imidazol, iminazole, 1,3-diaza-2,4-cyclopentadiene, glyoxaline, 1kg, 99% min. (GC), 23, 45, 255.0 deg.C to 256.0 deg.C, CHEBI:16069, White to Yellow or Beige, 206-019-2, 01,492; 02,220
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 301870025
Price (USD)/ Each
402.65
Online exclusive
447.00 
Save 44.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
402.65
Online exclusive
447.00 
Save 44.35 (10%)
Add to cart
Imidazole, 99+%, crystalline, Thermo Scientific Chemicals
Catalog number301870025
Price (USD)/ Each
402.65
Online exclusive
447.00 
Save 44.35 (10%)
-
Add to cart
Chemical Identifiers
CAS288-32-4
IUPAC Name1H-imidazole
Molecular FormulaC3H4N2
InChI KeyRAXXELZNTBOGNW-UHFFFAOYSA-N
SMILESN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
Appearance (Color)White to light yellow to light beige
Infrared spectrumConforms
Titration with HClO4>=99.0 % (On dry substance)
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only