Thermo Scientific Chemicals

2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95%, Thermo Scientific Chemicals

Catalog number: 302090100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

2-(3-Methoxyallylidene)malonic acid dimethyl ester, 95%, Thermo Scientific Chemicals

Catalog number: 302090100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 302090100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
41530-32-9
IUPAC Name
1,3-dimethyl 2-[(2E)-3-methoxyprop-2-en-1-ylidene]propanedioate
Molecular Formula
C9H12O5
InChI Key
NOBNTZJTOBIOHL-GQCTYLIASA-N
SMILES
CO\C=C\C=C(C(=O)OC)C(=O)OC
GC
>=94 %
Appearance (Form)
Low melting solid
Appearance (Color)
Clear yellow-green
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text