Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Thermo Scientific™

Samco™ UrineGARD™ Urine Collection Container

Green features
Prevent light exposure using tinted, Thermo Scientific™ Samco™ UrineGUARD™ Urine Collection Containers, which alleviate mess and exposure in urine collection and sampling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
302500
also known as 30 2500
2.5 L
303500
also known as 30 3500
3.5 L
Catalog number 302500
also known as 30 2500
Price (USD)
312.80
Online Exclusive
394.00
Save 81.20 (21%)
Case of 40
Add to cart
Volume (Metric):
2.5 L
Price (USD)
312.80
Online Exclusive
394.00
Save 81.20 (21%)
Case of 40
Add to cart

Prevent light exposure by using Thermo Scientific™ Samco™ UrineGUARD™ Urine Collection Containers. The Samco 24-hour Urine Collection product line alleviates mess and exposure when handling urine collection and sampling. Containers are inert to normal preservative.

  • 100% High Density Polyethylene (HDPE)
  • Large carrying handles for easy transportation
  • Easy-to-read graduations and highly accurate scales
  • Custom cap and mouth design helps prevent leakage
  • Upright configuration for compact storage
  • Custom spout allows for accurate pouring into test tubes, centrifuge tubes or aliquot bottles
Specifications
Closure TypeWide Mouth Screw Cap with Integral Spout
ColorAmber
Description24-hour UrineGUARD Specimen Container
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Green FeaturesLess waste
No. per Case40/Cs.
Volume (Metric)2.5 L
GraduatedYes
MaterialHDPE
Product TypeUrine Collection Container
Unit SizeCase of 40

Share catalog number, name or link