Thermo Scientific Chemicals

(+)-Dibenzoyl-D-tartaric acid, 98+%, Thermo Scientific Chemicals

Catalog number: 302511000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

(+)-Dibenzoyl-D-tartaric acid, 98+%, Thermo Scientific Chemicals

Catalog number: 302511000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 302511000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17026-42-5
IUPAC Name
(2S,3S)-2,3-bis(benzoyloxy)butanedioic acid
Molecular Formula
C18H14O8
InChI Key
YONLFQNRGZXBBF-ZCWZLOQUNA-N
SMILES
OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
HPLC
>=98.0 %
Appearance (Form)
Powder
Melting point
150°C to 155°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text