Thermo Scientific Chemicals

Linolenic acid, 99%

Catalog number: 302820010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Linolenic acid, 99%

Catalog number: 302820010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 302820010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
463-40-1
IUPAC Name
(9E,12E,15E)-octadeca-9,12,15-trienoic acid
Molecular Formula
C18H30O2
InChI Key
DTOSIQBPPRVQHS-IUQGRGSQSA-N
SMILES
CC\C=C\C\C=C\C\C=C\CCCCCCCC(O)=O
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4790 to 1.4810 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text