Linolenic acid, 99%
Thermo Scientific Chemicals

Linolenic acid, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 mg
1 g
Packaging:
Glass bottle
Catalog number 302820050
Price (USD)/ Each
443.65
Online exclusive
493.00 
Save 49.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
443.65
Online exclusive
493.00 
Save 49.35 (10%)
Add to cart
Linolenic acid, 99%
Catalog number302820050
Price (USD)/ Each
443.65
Online exclusive
493.00 
Save 49.35 (10%)
-
Add to cart
Chemical Identifiers
CAS463-40-1
IUPAC Name(9E,12E,15E)-octadeca-9,12,15-trienoic acid
Molecular FormulaC18H30O2
InChI KeyDTOSIQBPPRVQHS-IUQGRGSQSA-N
SMILESCC\C=C\C\C=C\C\C=C\CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4790 to 1.4810 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only