Thermo Scientific Chemicals

D-Arabitol, 99%

Catalog number: 302870250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

D-Arabitol, 99%

Catalog number: 302870250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 302870250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
488-82-4
IUPAC Name
pentane-1,2,3,4,5-pentol
Molecular Formula
C5H12O5
InChI Key
HEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)CO
Appearance (Color)
White to almost white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
100°C to 105°C
HPLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text