Thermo Scientific Chemicals

β-Alanine ethyleester hydrochloride, 98%

Catalog number: 303060500
50 g, Glass Bottle, Each
Thermo Scientific Chemicals

β-Alanine ethyleester hydrochloride, 98%

Catalog number: 303060500
50 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 303060500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4244-84-2
IUPAC Name
ethyl 3-aminopropanoate hydrochloride
Molecular Formula
C5H12ClNO2
InChI Key
RJCGNNHKSNIUAT-UHFFFAOYSA-N
SMILES
Cl.CCOC(=O)CCN
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
Titration Argentometric
97.5 to 102.5 %
TLC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text