Chemical Identifiers
CAS2492-87-7
IUPAC Name(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular FormulaC12H15NO8
InChI KeyIFBHRQDFSNCLOZ-RMPHRYRLSA-N
SMILESOC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
View more