Thermo Scientific Chemicals

4-Nitrophenyl-β-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Catalog number: 303110250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Nitrophenyl-β-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Catalog number: 303110250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 303110250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2492-87-7
IUPAC Name
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Formula
C12H15NO8
InChI Key
IFBHRQDFSNCLOZ-RMPHRYRLSA-N
SMILES
OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Appearance (Form)
Powder
HPLC
>=98.5 %
Specific optical rotation
-96° to -117° (20°C, 589 nm) (c=1, water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text