Chemical Identifiers
CAS105832-38-0
IUPAC Name[(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
Molecular FormulaC9H16BF4N3O3
InChI KeyYEBLHMRPZHNTEK-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
View more