Thermo Scientific Chemicals

4-Chloro-3-sulfamoylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 303220050
5 g, Each
Thermo Scientific Chemicals

4-Chloro-3-sulfamoylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 303220050
5 g, Each
Quantity
Catalog number: 303220050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1205-30-7
IUPAC Name
4-chloro-3-sulfamoylbenzoate
Molecular Formula
C7H5ClNO4S
InChI Key
FHQAWINGVCDTTG-UHFFFAOYSA-M
SMILES
NS(=O)(=O)C1=CC(=CC=C1Cl)C([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Melting point
258°C to 263°C
Appearance (Form)
Powder
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text