Thermo Scientific Chemicals

L-Lysine, 98%

Catalog number: 303341000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-Lysine, 98%

Catalog number: 303341000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 303341000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56-87-1
IUPAC Name
(2S)-2,6-diaminohexanoic acid
Molecular Formula
C6H14N2O2
InChI Key
KDXKERNSBIXSRK-MLHKIVSYNA-N
SMILES
NCCCC[C@H](N)C(O)=O
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 % (on dry substance)
Water
=<5.0 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text