Thermo Scientific Chemicals

4-Bromobutyryl chloride, 95%, Thermo Scientific Chemicals

Catalog number: 303440050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Bromobutyryl chloride, 95%, Thermo Scientific Chemicals

Catalog number: 303440050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 303440050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
927-58-2
IUPAC Name
4-bromobutanoyl chloride
Molecular Formula
C4H6BrClO
InChI Key
LRTRXDSAJLSRTG-UHFFFAOYSA-N
SMILES
ClC(=O)CCCBr
Appearance (Color)
Clear colorless to light yellow to light pink
Refractive index
1.4910 to 1.4940 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text