Thermo Scientific Chemicals

4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 303470100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 303470100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 303470100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15084-51-2
IUPAC Name
4-tert-butylbenzene-1-sulfonyl chloride
Molecular Formula
C10H13ClO2S
InChI Key
YEZADZMMVHWFIY-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC=C(C=C1)S(Cl)(=O)=O
GC
>=97.5 %
Appearance (Color)
White to off-white to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals and/or chunks
Melting point
78°C to 84°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text