Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Samco™ UrineGARD™ Urine Collection Container
Thermo Scientific™

Samco™ UrineGARD™ Urine Collection Container

Green features
Prevent light exposure using tinted, Thermo Scientific™ Samco™ UrineGUARD™ Urine Collection Containers, which alleviate mess and exposure in urine collection and sampling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
303500
also known as 30 3500
3.5 L
302500
also known as 30 2500
2.5 L
Catalog number 303500
also known as 30 3500
Price (USD)
174.60
Online Exclusive
194.00
Save 19.40 (10%)
Case of 24
Estimated availability date  Pending  
Add to cart
Volume (Metric):
3.5 L
Recurring order eligible. Learn more »
Price (USD)
174.60
Online Exclusive
194.00
Save 19.40 (10%)
Case of 24
Add to cart
Ask our AI about this Product

Prevent light exposure by using Thermo Scientific™ Samco™ UrineGUARD™ Urine Collection Containers. The Samco 24-hour Urine Collection product line alleviates mess and exposure when handling urine collection and sampling. Containers are inert to normal preservative.

  • 100% High Density Polyethylene (HDPE)
  • Large carrying handles for easy transportation
  • Easy-to-read graduations and highly accurate scales
  • Custom cap and mouth design helps prevent leakage
  • Upright configuration for compact storage
  • Custom spout allows for accurate pouring into test tubes, centrifuge tubes or aliquot bottles
Specifications
Closure TypeWide Mouth Screw Cap with Integral Spout
ColorAmber
Description24-hour UrineGUARD Specimen Container
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Green FeaturesLess waste
No. per Case24/Cs.
Volume (Metric)3.5 L
GraduatedYes
MaterialHDPE
Product TypeUrine Collection Container
Unit SizeCase of 24
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection