2,2,2-Trifluoroethylamine, 99.5%, Thermo Scientific Chemicals
2,2,2-Trifluoroethylamine, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,2-Trifluoroethylamine, 99.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Packaging:
Glass bottle
Catalog number 303500500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS753-90-2
IUPAC Name2,2,2-trifluoroethan-1-amine
Molecular FormulaC2H4F3N
InChI KeyKIPSRYDSZQRPEA-UHFFFAOYSA-N
SMILESNCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.4 %
Appearance (Color)Clear colorless to almost colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only