8-Methylquinoline, 97%, Thermo Scientific Chemicals
8-Methylquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Methylquinoline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 303620250
Price (USD)/ Each
201.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
201.00
Add to cart
8-Methylquinoline, 97%, Thermo Scientific Chemicals
Catalog number303620250
Price (USD)/ Each
201.00
-
Add to cart
Chemical Identifiers
CAS611-32-5
IUPAC Name8-methylquinoline
Molecular FormulaC10H9N
InChI KeyJRLTTZUODKEYDH-UHFFFAOYSA-N
SMILESCC1=C2N=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.6130 to 1.6170 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only