Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 303640050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 303640050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 303640050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
32247-96-4
IUPAC Name
1-(bromomethyl)-3,5-bis(trifluoromethyl)benzene
Molecular Formula
C9H5BrF6
InChI Key
ATLQGZVLWOURFU-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC(=CC(CBr)=C1)C(F)(F)F
Appearance (Color)
Clear colorless to yellow-brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=96.0 %
Refractive index
1.4440 to 1.4460 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text