Chemical Identifiers
CAS12141-67-2
IUPAC Namehexasodium tris(dioxotungstenbis(olate)) nonakis(trioxotungsten)
Molecular FormulaNa6O39W12
InChI KeyAZCSOJKJFMWYCX-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O
View more