Thermo Scientific Chemicals

Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 304170050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 304170050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 304170050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
252976-51-5
IUPAC Name
ytterbium(3+) tritrifluoromethanesulfonate
Molecular Formula
C3F9O9S3Yb
InChI Key
AHZJKOKFZJYCLG-UHFFFAOYSA-K
SMILES
[Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Total conc. of imp.as REO
=<0.1 %
Ytterbium (Yb)
20.0 to 26.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text