Thermo Scientific Chemicals

2-chloro-4-hydroxybenzoic acid hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 304200010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-chloro-4-hydroxybenzoic acid hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 304200010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 304200010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56363-84-9
IUPAC Name
2-chloro-4-hydroxybenzoic acid
Molecular Formula
C7H5ClO3
InChI Key
WIPYZRZPNMUSER-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O)C=C1Cl
Appearance (Color)
Off-white to beige to orange-brown
Infrared spectrum
Conforms
Water
=<8 % (K.F.)
Appearance (Form)
Crystalline powder
Titration with TBAH
>=97.5 % (On dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text